Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022712.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022712.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022712.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022712.hg.1 |
TC12001703.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022711.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022711.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12022711.hg.1 |
TC12001703.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000433116.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000433116.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000433116.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186846.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186846.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186846.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186846.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186846.hg.1 |
TC1200011195.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200186849.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186849.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186849.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186849.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186849.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186849.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186849.hg.1 |
TC1200011195.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200186847.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186847.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186847.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186847.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186847.hg.1 |
TC1200011195.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000433116.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000433116.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000433116.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011195.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186848.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186848.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186848.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186848.hg.1 |
ENST00000500695.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186848.hg.1 |
ENST00000502102.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200186848.hg.1 |
NR_038930.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200186848.hg.1 |
TC1200011195.hg.1 |
Clariom D TC |