Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
230631_PM_s_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
227125_PM_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
230632_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004604.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004604.hg.1 |
NR_038974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004604.hg.1 |
TC21000408.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004605.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004605.hg.1 |
NR_038974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21004605.hg.1 |
TC21000408.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007986.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007986.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007986.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007986.hg.1 |
NR_038974.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007986.hg.1 |
NR_038974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153811.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153811.hg.1 |
NR_038974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153811.hg.1 |
TC2100007986.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153811.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153811.hg.1 |
NR_038974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153810.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153810.hg.1 |
NR_038974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100153810.hg.1 |
TC2100007986.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100153810.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100153810.hg.1 |
NR_038974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007986.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007986.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007986.hg.1 |
ENST00000411998.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007986.hg.1 |
NR_038974.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007986.hg.1 |
NR_038974.1 |
Refseq |
Human Genome U133 Set |
230632_at |
- |
- |
Human Genome U133 Set |
230631_s_at |
- |
- |
Human Genome U133 Set |
230631_PM_s_at |
- |
- |
Human Genome U133 Set |
230632_PM_at |
- |
- |