Assay Details
| Gene Symbol: MIR4422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039618.1 | 1 | 38 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100507634 | LOC100507634 | -8.2kb | Sense | Chr. 1 - 55215408 - 55217455 on NCBI Build GRCh38 | |
| Downstream | phospholipid phosphatase 3 | PLPP3 |
Dri42, LPP3, PAP2B, PPAP2B, VCIP |
1.3mb | Anti-sense | Chr. 1 - 56494747 - 56579584 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0100008423.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000678.hg.1
Set Membership: |
|

MIR4422
Genomic Map