Assay Details
Gene Symbol: MIR4425 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_039623.1 | 1 | 43 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | runt related transcription factor 3 | RUNX3 |
AML2, CBFA3, PEBP2aC |
-58.3kb | Anti-sense | Chr. 1 - 24899511 - 24965158 on NCBI Build GRCh38 |
Downstream | SYF2 pre-mRNA splicing factor | SYF2 |
CBPIN, NTC31, P29, fSAP29 |
198.7kb | Anti-sense | Chr. 1 - 25222276 - 25232522 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007384.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
