Assay Details
| Gene Symbol: MIR4653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039797.1 | 1 | 46 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serpin family E member 1 | SERPINE1 |
PAI, PAI-1, PAI1, PLANH1 |
-20.2kb | Sense | Chr. 7 - 101127087 - 101139266 on NCBI Build GRCh38 |
| Downstream | VGF nerve growth factor inducible | VGF |
SCG7, SgVII |
3.0kb | Anti-sense | Chr. 7 - 101162509 - 101169956 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as Clariom D transcript cluster(s) TC0700008585.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000645.hg.1
Set Membership: |
|

MIR4653
Genomic Map