Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011675.hg.1 |
ENST00000582039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011675.hg.1 |
NR_039808.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020702.hg.1 |
TC08001580.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08020702.hg.1 |
NR_039808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179198.hg.1 |
ENST00000582039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179198.hg.1 |
NR_039808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179198.hg.1 |
ENST00000582039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800179198.hg.1 |
NR_039808.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800179198.hg.1 |
TC0800011675.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800011675.hg.1 |
ENST00000582039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011675.hg.1 |
NR_039808.1 |
Refseq |