Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007222.hg.1 |
ENST00000582691.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007222.hg.1 |
NR_039863.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003756.hg.1 |
NR_039863.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003756.hg.1 |
TC15000403.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500151526.hg.1 |
ENST00000582691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151526.hg.1 |
NR_039863.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151526.hg.1 |
ENST00000582691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500151526.hg.1 |
NR_039863.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500151526.hg.1 |
TC1500007222.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007222.hg.1 |
ENST00000582691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007222.hg.1 |
NR_039863.1 |
Refseq |