| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001723.hg.1 |
ENST00000501265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001723.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001723.hg.1 |
TC13000149.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001722.hg.1 |
ENST00000501265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001722.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001722.hg.1 |
TC13000149.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
ENST00000612345.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
ENST00000612345.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
ENST00000612345.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
ENST00000612345.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147374.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147374.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147374.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147374.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147374.hg.1 |
TC1300006985.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300147375.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147375.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147375.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147375.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147375.hg.1 |
TC1300006985.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
ENST00000612345.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
ENST00000612345.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
ENST00000612345.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
ENST00000612345.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006985.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147376.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147376.hg.1 |
ENST00000612345.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147376.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147376.hg.1 |
ENST00000611103.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147376.hg.1 |
ENST00000612345.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147376.hg.1 |
NR_039974.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147376.hg.1 |
TC1300006985.hg.1 |
Clariom D TC |