Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026241.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17026241.hg.1 |
TC17001926.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
ENST00000581579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
ENST00000581579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
ENST00000581579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
ENST00000581579.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011926.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
ENST00000581579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
ENST00000581579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
ENST00000581579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
ENST00000581579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011926.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197494.hg.1 |
ENST00000581579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197494.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197494.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197494.hg.1 |
ENST00000581579.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197494.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197494.hg.1 |
NR_040019.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197494.hg.1 |
TC1700011926.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700197496.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197496.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197496.hg.1 |
ENST00000577521.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700197496.hg.1 |
NR_040018.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700197496.hg.1 |
TC1700011926.hg.1 |
Clariom D TC |