Assay Details
| Gene Symbol: IQCH-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040051.1 | 4 - 5 | 930 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | alpha- and gamma-adaptin binding protein | AAGAB |
KPPP1, PPKP1, PPKP1A, p34 |
-166.1kb | Sense | Chr. 15 - 67200675 - 67255198 on NCBI Build GRCh38 |
| Downstream | chromosome 15 open reading frame 61 | C15orf61 | 61.2kb | Anti-sense | Chr. 15 - 67521184 - 67527303 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001588.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15015295.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015295.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015295.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015295.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015295.hg.1 | TC15001588.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015296.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015296.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015296.hg.1 | TC15001588.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015292.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015292.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015292.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015292.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015292.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015292.hg.1 | TC15001588.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040055.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009848.hg.1 | NR_040056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015294.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015294.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015294.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15015294.hg.1 | TC15001588.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000558025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000559702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040054.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040055.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009848.hg.1 | NR_040056.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173056.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173056.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173056.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173056.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173056.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173056.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173056.hg.1 | TC1500009848.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500173054.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173054.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173054.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173054.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173054.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173054.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173054.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173054.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173054.hg.1 | TC1500009848.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | ENST00000559285.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | ENST00000559298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | ENST00000561232.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | NR_040051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | NR_040052.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500173051.hg.1 | TC1500009848.hg.1 | Clariom D TC |

IQCH-AS1
Genomic Map