Assay Details
Gene Symbol: LINC01630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040074.1 | 1 - 2 | 218 | 81 | ||
GenBank mRNA | AK294844.1 | - | 1 - 2 | 218 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mex-3 RNA binding family member C | MEX3C |
BM-013, MEX-3C, RKHD2, RNF194 |
-194.4kb | Anti-sense | Chr. 18 - 51174550 - 51197681 on NCBI Build GRCh38 |
Downstream | DCC netrin 1 receptor | DCC |
CRC18, CRCR1, IGDCC1, MRMV1, NTN1R1 |
779.3kb | Sense | Chr. 18 - 52340172 - 53535903 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000182.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK294844.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18002150.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18002150.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18002150.hg.1 | TC18000182.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000578152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000578152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000578152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000578152.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | NR_040075.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007335.hg.1 | NR_040075.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | TC1800007335.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150220.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | TC1800007335.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150219.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150221.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150221.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150221.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150221.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800150221.hg.1 | TC1800007335.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800150221.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150221.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150221.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800150221.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000435144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000578152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000578152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000578152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000578152.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000580841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000582689.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583609.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | ENST00000583982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | NR_040074.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | NR_040075.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007335.hg.1 | NR_040075.1 | Refseq |
