Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
ENST00000415147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
ENST00000415147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008063.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005073.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005073.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005073.hg.1 |
TC21000429.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005072.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005072.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005072.hg.1 |
TC21000429.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100154465.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154465.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154465.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154465.hg.1 |
TC2100008063.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154465.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154465.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154465.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
ENST00000415147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
ENST00000415147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008063.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154466.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154466.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154466.hg.1 |
NR_040084.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154466.hg.1 |
TC2100008063.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154466.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154466.hg.1 |
ENST00000535199.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154466.hg.1 |
NR_040084.1 |
Refseq |