Assay Details
Gene Symbol: LOC392364 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040117.1 | 1 | 919 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC497256 | LOC497256 | -10.3kb | Sense | Chr. 9 - 87844517 - 87939244 on NCBI Build GRCh38 | |
Downstream | SPATA31 subfamily E member 1 | SPATA31E1 |
C9orf79, FAM75E1 |
23.8kb | Anti-sense | Chr. 9 - 87882857 - 87888899 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001302.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16236702 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06801034 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06801018 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141043 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141057 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141030 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141053 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141060 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16236705 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141040 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141031 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141061 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141046 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141033 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06801000 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141039 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06801007 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16236704 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06800992 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06800997 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06801025 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141035 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06801022 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16236711 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141020 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141051 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141048 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16236710 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06801030 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06800996 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141050 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141042 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141055 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141037 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16236709 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06801011 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16236701 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141049 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141054 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06800994 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141052 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P27141045 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06800993 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06801001 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 19278 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 46726 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3293082 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3375762 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8162192 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8162193 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8162191 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR09016293.hg.1 | TC09001302.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09016293.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09016292.hg.1 | TC09001302.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09016292.hg.1 | ENST00000399186.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09016292.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09016294.hg.1 | TC09001302.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09016294.hg.1 | ENST00000399186.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09016294.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172705.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172705.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172705.hg.1 | TC0900010658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900172706.hg.1 | ENST00000399186.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172706.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172706.hg.1 | ENST00000399186.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172706.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172706.hg.1 | TC0900010658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900172707.hg.1 | ENST00000399186.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172707.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172707.hg.1 | ENST00000399186.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172707.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172707.hg.1 | TC0900010658.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900172704.hg.1 | ENST00000399186.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172704.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172704.hg.1 | ENST00000399186.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172704.hg.1 | NR_040117.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172704.hg.1 | TC0900010658.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1730590 | BC086877.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1730590 | NR_040117.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1750317 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1750317 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1730590 | BC086877.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1730590 | NR_040117.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1730590 | BC086877.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1730590 | NR_040117.1 | human_transcriptome |
