Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011801.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025045.hg.1 |
ENST00000551505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025045.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025045.hg.1 |
TC12001920.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025042.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025042.hg.1 |
TC12001920.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025043.hg.1 |
ENST00000551505.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025043.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025043.hg.1 |
TC12001920.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025044.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025044.hg.1 |
TC12001920.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011801.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191464.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191464.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191464.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191464.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191464.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191464.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191464.hg.1 |
TC1200011801.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200191466.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191466.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191466.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191466.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191466.hg.1 |
ENST00000551505.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191466.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191466.hg.1 |
TC1200011801.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200191465.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191465.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191465.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191465.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191465.hg.1 |
TC1200011801.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200191463.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191463.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191463.hg.1 |
ENST00000549203.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191463.hg.1 |
NR_040246.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191463.hg.1 |
TC1200011801.hg.1 |
Clariom D TC |