Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008945.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008945.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008945.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008945.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008945.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008945.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008945.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008945.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008945.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014710.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014710.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22014710.hg.1 |
TC22000868.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200164892.hg.1 |
TC2200008945.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200164892.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164892.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200164892.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200164892.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008945.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008945.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008945.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008945.hg.1 |
ENST00000443783.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008945.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008945.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008945.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008945.hg.1 |
NR_044991.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008945.hg.1 |
NR_044991.1 |
Refseq |