Assay Details
| Gene Symbol: SNHG18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045196.2 | 1 - 2 | 326 | 62 | ||
| GenBank mRNA | DA542304.1 | - | 1 - 2 | 330 | 62 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000066.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05000743.hg.1 | TC05000066.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000743.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000743.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000743.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000746.hg.1 | TC05000066.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000746.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000746.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000746.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000744.hg.1 | TC05000066.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000744.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000744.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000744.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000459473.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | NR_003689.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000747.hg.1 | TC05000066.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000747.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000747.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000742.hg.1 | TC05000066.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000742.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05000742.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145955.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145955.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145955.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145955.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145955.hg.1 | TC0500006712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500145944.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145944.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145944.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145944.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145944.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145944.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145944.hg.1 | TC0500006712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500145945.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145945.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145945.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145945.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145945.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145945.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145945.hg.1 | TC0500006712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500145954.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145954.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145954.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145954.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145954.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145954.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145954.hg.1 | TC0500006712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500145956.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145956.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145956.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145956.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145956.hg.1 | TC0500006712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000459473.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | NR_003689.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006712.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145943.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145943.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145943.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145943.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145943.hg.1 | TC0500006712.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500145951.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145951.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145951.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145951.hg.1 | ENST00000508179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145951.hg.1 | ENST00000509788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500145951.hg.1 | NR_045196.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500145951.hg.1 | TC0500006712.hg.1 | Clariom D TC |

SNHG18
Genomic Map