Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003657.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003657.hg.1 |
TC19000253.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003659.hg.1 |
ENST00000498856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003659.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003659.hg.1 |
TC19000253.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003658.hg.1 |
ENST00000498856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003658.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003658.hg.1 |
TC19000253.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
ENST00000588658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
ENST00000588658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
ENST00000588658.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
ENST00000592086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
ENST00000592086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007160.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900151658.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151658.hg.1 |
ENST00000588658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151658.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900151658.hg.1 |
TC1900007160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900151658.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151658.hg.1 |
ENST00000588658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151658.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900151660.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151660.hg.1 |
ENST00000588658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151660.hg.1 |
ENST00000592086.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151660.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900151660.hg.1 |
TC1900007160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900151660.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151660.hg.1 |
ENST00000588658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151660.hg.1 |
ENST00000592086.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900151660.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900151656.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900151656.hg.1 |
TC1900007160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900151656.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
ENST00000588387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
ENST00000588658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
ENST00000588658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
ENST00000588658.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
ENST00000592086.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
ENST00000592086.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
NR_045214.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007160.hg.1 |
NR_045214.1 |
Refseq |