Assay Details
| Gene Symbol: TRIM41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045218.1 | 3 - 4 | 1874 | 117 | ||
| GenBank mRNA | BC004956.1 | - | 3 - 4 | 1079 | 117 | |
| BC071887.1 | - | 3 - 4 | 1079 | 117 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4638 | MIR4638 | mir-4638 | -10.0kb | Anti-sense | Chr. 5 - 181222566 - 181222633 on NCBI Build GRCh38 |
| Downstream | receptor for activated C kinase 1 | RACK1 |
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21 |
3.2kb | Anti-sense | Chr. 5 - 181236928 - 181243906 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

TRIM41
Genomic Map