Assay Details
| Gene Symbol: SRGAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC150646.1 | - | 3 - 4 | 920 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 72 member A | FAM72A |
LMPIP, Ugene |
-137.9kb | Anti-sense | Chr. 1 - 206186179 - 206204918 on NCBI Build GRCh38 |
| Downstream |
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon |
IKBKE |
IKK-E, IKK-i, IKKE, IKKI |
86.1kb | Sense | Chr. 1 - 206470243 - 206496890 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SRGAP2
Genomic Map