Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025135.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12025135.hg.1 |
TC12001931.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011828.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011828.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011828.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011828.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011828.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011828.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011828.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191646.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191646.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191646.hg.1 |
TC1200011828.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200191648.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191648.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191648.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191648.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191648.hg.1 |
TC1200011828.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200191649.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191649.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191649.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191649.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191649.hg.1 |
TC1200011828.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200011828.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011828.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011828.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011828.hg.1 |
ENST00000547904.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011828.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011828.hg.1 |
NR_037629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011828.hg.1 |
NR_037629.1 |
Refseq |