Assay Details
| Gene Symbol: NBAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034143.1 | 3 | 924 | 120 | ||
| GenBank mRNA | AK126168.1 | - | 3 | 924 | 120 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cancer susceptibility candidate 15 (non-protein coding) | CASC15 | LINC00340 | Anti-sense | Chr. 6 - 21666444 - 22194400 on NCBI Build GRCh38 | |
| Downstream | prolactin | PRL | GHA1 | 150.6kb | Sense | Chr. 6 - 22287244 - 22302897 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001319.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605606 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794100 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794081 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794077 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24793966 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605632 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794062 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605576 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794122 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605579 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605561 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605474 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794086 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794095 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605581 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794126 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794110 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794063 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794116 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24793969 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605552 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794098 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24793968 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605557 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24793967 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794092 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605562 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605590 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794052 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794101 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794089 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605478 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794115 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605556 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605565 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794070 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605560 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605619 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605617 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605558 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794130 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605570 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794056 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794102 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794087 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605605 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605612 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605597 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605554 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24793974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24793971 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794064 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605634 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794118 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794065 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605591 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794061 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794097 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794106 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15791496 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24793972 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794114 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04605623 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794090 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794080 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794076 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24794091 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3214066 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 20470 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3214066 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3214066 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 241670_PM_x_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015751.hg.1 | TC06001319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015751.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015752.hg.1 | TC06001319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015752.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015750.hg.1 | TC06001319.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06015750.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174684.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174684.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174684.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174684.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174684.hg.1 | TC0600011031.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600174688.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174688.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174688.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174688.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174688.hg.1 | TC0600011031.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011031.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174683.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174683.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174683.hg.1 | TC0600011031.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600174685.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174685.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174685.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174685.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174685.hg.1 | TC0600011031.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600174682.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174682.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174682.hg.1 | TC0600011031.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600174686.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174686.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174686.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174686.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174686.hg.1 | TC0600011031.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600174689.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174689.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174689.hg.1 | ENST00000566912.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600174689.hg.1 | NR_034143.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600174689.hg.1 | TC0600011031.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 241670_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 241670_PM_x_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 241670_x_at6 | - | - |
| Human Genome U133 Set | 241670_PM_x_at | - | - |
| Human Genome U133 Set | 241670_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029472 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029472 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3303979 | AK126168.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3303979 | NR_034143.1 | human_transcriptome |

NBAT1
Genomic Map