Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009270.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009270.hg.1 |
TC10000778.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008790.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009272.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009272.hg.1 |
TC10000778.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009271.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009271.hg.1 |
TC10000778.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000160140.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160140.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160140.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160140.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160140.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160140.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160140.hg.1 |
TC1000008790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000160142.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160142.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160142.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160142.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160142.hg.1 |
TC1000008790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008790.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160141.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160141.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160141.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160141.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160141.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160141.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160141.hg.1 |
TC1000008790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000160138.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160138.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160138.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160138.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160138.hg.1 |
TC1000008790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000160139.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160139.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160139.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160139.hg.1 |
ENST00000625265.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160139.hg.1 |
ENST00000627886.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160139.hg.1 |
NR_038940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160139.hg.1 |
TC1000008790.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
1557389_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1557389_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557389_at2 |
- |
- |