Assay Details
Gene Symbol: GAS6-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_044993.1 | 2 | 1221 | 87 | ||
GenBank mRNA | BX109451.1 | - | 1 | 169 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | growth arrest specific 6 | GAS6 |
AXLLG, AXSF |
-907.0 | Anti-sense | Chr. 13 - 113820549 - 113864103 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 454 | LINC00454 | 11.8kb | Anti-sense | Chr. 13 - 113878628 - 113883378 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000431.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13004893.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13004893.hg.1 | TC13000431.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13004894.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13004894.hg.1 | TC13000431.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000420382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000420382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154928.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154928.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154928.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154928.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154928.hg.1 | TC1300008153.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300154926.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154926.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154926.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154926.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154926.hg.1 | TC1300008153.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300154927.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154927.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154927.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154927.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154927.hg.1 | TC1300008153.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300154929.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154929.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154929.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154929.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154929.hg.1 | TC1300008153.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300154924.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154924.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154924.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154924.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154924.hg.1 | TC1300008153.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300154930.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154930.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154930.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300154930.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300154930.hg.1 | TC1300008153.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000420382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000420382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | ENST00000608651.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008153.hg.1 | NR_044993.1 | Refseq |
Human Genome U95 Set | 74731_at | - | - |
Human Genome U95 Set | 74731_PM_at | - | - |
