| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023286.hg.1 |
TC03003407.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023286.hg.1 |
ENST00000455218.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023286.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023286.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013965.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013965.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013965.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013965.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013965.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013965.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013965.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013965.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013965.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300013965.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023285.hg.1 |
TC03003407.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023285.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023285.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023287.hg.1 |
TC03003407.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023287.hg.1 |
ENST00000455218.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023287.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03023287.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013965.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013965.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013965.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013965.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013965.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300013965.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013965.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013965.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013965.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300013965.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300207242.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300207242.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300207242.hg.1 |
ENST00000463091.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300207242.hg.1 |
NM_001205271.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300207242.hg.1 |
TC0300013965.hg.1 |
Clariom D TC |