Assay Details
Gene Symbol: ZFP36L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244698.1 | NP_001231627.1 | 2 - 3 | 1992 | 85 | |
GenBank mRNA | AK024202.1 | - | 2 - 3 | 2816 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14001253.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
vasculogenesis
proepicardium development
regulation of transcription, DNA-templated
apoptotic process
heart development
cell proliferation
response to wounding
regulation of gene expression
regulation of keratinocyte proliferation
phosphatidylinositol 3-kinase signaling
neural tube development
nuclear-transcribed mRNA catabolic process, deadenylation-independent decay
regulation of mRNA 3'-end processing
cellular response to insulin stimulus
T cell differentiation in thymus
multicellular organism growth
p38MAPK cascade
regulation of mRNA stability
protein kinase B signaling
cellular response to fibroblast growth factor stimulus
positive regulation of fat cell differentiation
regulation of keratinocyte differentiation
negative regulation of erythrocyte differentiation
positive regulation of monocyte differentiation
regulation of myoblast differentiation
mesendoderm development
mRNA transport
chorio-allantoic fusion
spongiotrophoblast layer development
3'-UTR-mediated mRNA destabilization
ERK1 and ERK2 cascade
cellular response to cAMP
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
cellular response to peptide hormone stimulus
cellular response to glucocorticoid stimulus
cellular response to hypoxia
cellular response to salt stress
cellular response to transforming growth factor beta stimulus
regulation of stem cell proliferation
cellular response to raffinose
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
regulation of keratinocyte apoptotic process
positive regulation of intracellular mRNA localization
cellular response to phorbol 13-acetate 12-myristate
DNA binding
transcription factor activity, sequence-specific DNA binding
mRNA binding
mRNA 3'-UTR binding
protein binding
mRNA 3'-UTR AU-rich region binding
poly(A) RNA binding
metal ion binding
14-3-3 protein binding
MAPK cascade
nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
vasculogenesis
proepicardium development
regulation of transcription, DNA-templated
apoptotic process
heart development
cell proliferation
response to wounding
regulation of gene expression
regulation of keratinocyte proliferation
phosphatidylinositol 3-kinase signaling
neural tube development
nuclear-transcribed mRNA catabolic process, deadenylation-independent decay
regulation of mRNA 3'-end processing
cellular response to insulin stimulus
T cell differentiation in thymus
multicellular organism growth
p38MAPK cascade
regulation of mRNA stability
protein kinase B signaling
cellular response to fibroblast growth factor stimulus
positive regulation of fat cell differentiation
regulation of keratinocyte differentiation
negative regulation of erythrocyte differentiation
positive regulation of monocyte differentiation
regulation of myoblast differentiation
mesendoderm development
mRNA transport
chorio-allantoic fusion
spongiotrophoblast layer development
3'-UTR-mediated mRNA destabilization
ERK1 and ERK2 cascade
cellular response to cAMP
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
cellular response to peptide hormone stimulus
cellular response to glucocorticoid stimulus
cellular response to hypoxia
cellular response to salt stress
cellular response to transforming growth factor beta stimulus
regulation of stem cell proliferation
cellular response to raffinose
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
regulation of keratinocyte apoptotic process
positive regulation of intracellular mRNA localization
cellular response to phorbol 13-acetate 12-myristate
DNA binding
transcription factor activity, sequence-specific DNA binding
mRNA binding
mRNA 3'-UTR binding
protein binding
mRNA 3'-UTR AU-rich region binding
poly(A) RNA binding
metal ion binding
14-3-3 protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR14014221.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14014221.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14014221.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14014221.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14014221.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14014221.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14014221.hg.1 | TC14001253.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000553375.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557022.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | ENST00000557086.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009524.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170219.hg.1 | TC1400009524.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | ENST00000336440.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | ENST00000439696.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | ENST00000555997.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | NM_001244698.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | NM_001244701.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | NM_004926.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400170216.hg.1 | TC1400009524.hg.1 | Clariom D TC |
