Assay Details
| Gene Symbol: TAOK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252043.1 | NP_001238972.1 | 15 - 16 | 2804 | 101 | |
| NM_004783.3 | NP_004774.1 | 15 - 16 | 2804 | 101 | ||
| NM_016151.3 | NP_057235.2 | 15 - 16 | 2804 | 101 | ||
| XM_011545982.1 | XP_011544284.1 | 15 - 16 | 3051 | 101 | ||
| XM_011545983.2 | XP_011544285.1 | 15 - 16 | 3048 | 101 | ||
| XM_011545984.1 | XP_011544286.1 | 15 - 16 | 3051 | 101 | ||
| XM_011545985.1 | XP_011544287.1 | 15 - 16 | 3048 | 101 | ||
| XM_011545986.2 | XP_011544288.1 | 15 - 16 | 3051 | 101 | ||
| GenBank mRNA | AB020688.1 | - | 15 - 16 | 2390 | 101 | |
| AF061943.1 | - | 15 - 16 | 2745 | 101 | ||
| AF263313.1 | - | 14 - 15 | 1987 | 101 | ||
| AK291473.1 | - | 15 - 16 | 2321 | 101 | ||
| AK304446.1 | - | 7 - 8 | 1310 | 101 | ||
| AL137701.1 | - | 7 - 8 | 2641 | 101 | ||
| BC015204.1 | - | 15 - 16 | 2770 | 101 | ||
| BC051798.1 | - | 15 - 16 | 2561 | 101 | ||
| BC054495.1 | - | 15 - 16 | 2757 | 101 | ||
| BC136653.1 | - | 15 - 16 | 2489 | 9020643 | 101 | |
| BC136655.1 | - | 15 - 16 | 2489 | 9020645 | 101 | |
| BC142663.1 | - | 15 - 16 | 2144 | 40148112 | 101 | |
| BC144344.1 | - | 15 - 16 | 2489 | 9052864 | 101 | |
| BC151221.1 | - | 15 - 16 | 2144 | 40148752 | 101 | |
| BC152413.1 | - | 15 - 16 | 2390 | 8862518 | 101 | |
More Information
Set Membership: |
|

TAOK2
Genomic Map