Assay Details
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018073.7 | NP_060543.5 | 2 - 3 | 722 | 100 | |
GenBank mRNA | AF360739.1 | - | 2 - 3 | 675 | 100 | |
AF439153.1 | - | 1 - 2 | 432 | 100 | ||
AK056523.1 | - | 2 - 3 | 684 | 100 | ||
AK291166.1 | - | 2 - 3 | 634 | 100 | ||
AK310888.1 | - | 2 - 3 | 631 | 100 | ||
AL832730.1 | - | 2 - 3 | 672 | 100 | ||
AY005802.1 | - | 1 - 2 | 108 | 100 | ||
BC075058.2 | - | 1 - 2 | 456 | 30915367 | 100 | |
BC109063.2 | - | 2 - 3 | 641 | 40033795 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001311.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11016676.hg.1 | ENST00000300747.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11016676.hg.1 | ENST00000544055.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11016676.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11016676.hg.1 | TC11001311.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000531717.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | ENST00000533021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009884.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | ENST00000300747.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | ENST00000526337.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | ENST00000531101.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | ENST00000531644.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | ENST00000532108.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | NM_001304496.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | NM_018073.7 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100174755.hg.1 | TC1100009884.hg.1 | Clariom D TC |
