Assay Details
| Gene Symbol: C2orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213608.2 | NP_998773.1 | 2 - 3 | 1264 | 96 | |
| GenBank mRNA | AY358249.1 | - | 2 - 3 | 1264 | 96 | |
| BC137150.1 | - | 2 - 3 | 1140 | 9021147 | 96 | |
| BC137151.1 | - | 2 - 3 | 1140 | 9021148 | 96 | |
More Information
Set Membership: |
|

C2orf66
Genomic Map