Assay Details
| Gene Symbol: IKZF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257408.1 | NP_001244337.1 | 1 - 2 | 61 | 89 | |
| NM_001257409.1 | NP_001244338.1 | 1 - 2 | 61 | 89 | ||
| NM_001257410.1 | NP_001244339.1 | 1 - 2 | 61 | 89 | ||
| NM_001257411.1 | NP_001244340.1 | 1 - 2 | 61 | 89 | ||
| NM_001257412.1 | NP_001244341.1 | 1 - 2 | 61 | 89 | ||
| NM_001257413.1 | NP_001244342.1 | 1 - 2 | 61 | 89 | ||
| NM_001257414.1 | NP_001244343.1 | 1 - 2 | 61 | 89 | ||
| NM_001284514.1 | NP_001271443.1 | 1 - 2 | 61 | 89 | ||
| NM_001284515.1 | NP_001271444.1 | 1 - 2 | 61 | 89 | ||
| NM_012481.4 | NP_036613.2 | 1 - 2 | 61 | 89 | ||
| NM_183228.2 | NP_899051.1 | 1 - 2 | 61 | 89 | ||
| NM_183229.2 | NP_899052.1 | 1 - 2 | 61 | 89 | ||
| NM_183230.2 | NP_899053.1 | 1 - 2 | 61 | 89 | ||
| NM_183231.2 | NP_899054.1 | 1 - 2 | 61 | 89 | ||
| NM_183232.2 | NP_899055.1 | 1 - 2 | 61 | 89 | ||
| GenBank mRNA | AY377973.1 | - | 1 - 2 | 61 | 89 | |
| AY377975.1 | - | 1 - 2 | 61 | 89 | ||
| AY377976.1 | - | 1 - 2 | 61 | 89 | ||
| AY377977.1 | - | 1 - 2 | 61 | 89 | ||
| AY377978.1 | - | 1 - 2 | 61 | 89 | ||
| AY377979.1 | - | 1 - 2 | 61 | 89 | ||
| AY377980.1 | - | 1 - 2 | 61 | 89 | ||
| AY377981.1 | - | 1 - 2 | 61 | 89 | ||
| AY377982.1 | - | 1 - 2 | 61 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY377973.1, AY377975.1, AY377976.1, AY377977.1, AY377978.1, AY377979.1, AY377980.1, AY377981.1, AY377982.1, NM_001257408.1, NM_001257409.1, NM_001257410.1, NM_001257411.1, NM_001257412.1, NM_001257413.1, NM_001257414.1, NM_001284514.1, NM_001284515.1, NM_012481.4, NM_183228.2, NM_183229.2, NM_183230.2, NM_183231.2, NM_183232.2 transcripts.
Set Membership: |
|

IKZF3
Genomic Map