Assay Details
Gene Symbol: IGLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178126.1 | NP_001171597.1 | 1 - 2 | 477 | 133 | |
GenBank mRNA | AJ318022.1 | - | 1 - 2 | 477 | 133 | |
BG756003.1 | - | 1 - 2 | 421 | 133 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22001450.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000385097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000385097.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390319.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390319.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390319.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390320.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390320.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390321.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390321.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000390323.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000522922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000526893.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000531372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000531372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000531372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000531372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000532223.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000577998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000610541.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000612695.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000613777.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000616131.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617866.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000617880.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620657.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | ENST00000620665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001178126.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001256296.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001256296.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001256296.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001256296.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NM_001256296.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009219.hg.1 | NR_049835.1 | Refseq |
