Assay Details
Gene Symbol: HCG22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003948.2 | 2 - 3 | 499 | 112 | ||
GenBank mRNA | AB600644.1 | - | 2 - 3 | 499 | 112 | |
AK094433.1 | - | 1 - 2 | 310 | 112 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mucin 22 | MUC22 | PBMUCL1 | -18.5kb | Sense | Chr. 6 - 31005952 - 31035402 on NCBI Build GRCh38 |
Downstream | chromosome 6 open reading frame 15 | C6orf15 | STG | 54.3kb | Anti-sense | Chr. 6 - 31111223 - 31112555 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000355.hg.1, TC6_apd_hap1000033.hg.1, TC6_cox_hap2000053.hg.1, TC6_dbb_hap3000046.hg.1, TC6_mann_hap4000047.hg.1, TC6_mcf_hap5000040.hg.1, TC6_qbl_hap6000046.hg.1, TC6_ssto_hap7000041.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000654.hg.1 | TC6_mcf_hap5000040.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000654.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000790.hg.1 | TC6_qbl_hap6000046.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000790.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000566475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000566475.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000570223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000570223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000361.hg.1 | TC6_apd_hap1000033.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000361.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000604.hg.1 | TC6_ssto_hap7000041.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000604.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000791.hg.1 | TC6_qbl_hap6000046.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6000791.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003391.hg.1 | TC06000355.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06003391.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06003391.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06003392.hg.1 | TC06000355.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06003392.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000776.hg.1 | TC6_mann_hap4000047.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000776.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000652.hg.1 | TC6_mcf_hap5000040.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000652.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000781.hg.1 | TC6_dbb_hap3000046.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000781.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000651.hg.1 | TC6_mcf_hap5000040.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5000651.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000360.hg.1 | TC6_apd_hap1000033.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000360.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000799.hg.1 | TC6_cox_hap2000053.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000799.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000780.hg.1 | TC6_dbb_hap3000046.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3000780.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000798.hg.1 | TC6_cox_hap2000053.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2000798.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000775.hg.1 | TC6_mann_hap4000047.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4000775.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000603.hg.1 | TC6_ssto_hap7000041.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7000603.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151517.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151517.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151517.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151517.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151517.hg.1 | TC0600007559.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151529.hg.1 | TC0600007559.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000566475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000566475.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000570223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000570223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007559.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151521.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151521.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151521.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151521.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151521.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151521.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151521.hg.1 | TC0600007559.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600151526.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151526.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151526.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151526.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151526.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151526.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151526.hg.1 | TC0600007559.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600151519.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151519.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151519.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151519.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151519.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151519.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151519.hg.1 | TC0600007559.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | ENST00000566475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | ENST00000570223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | ENST00000565192.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | ENST00000566475.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | ENST00000570223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | ENST00000615046.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151523.hg.1 | TC0600007559.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600151527.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151527.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151527.hg.1 | ENST00000570223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151527.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151527.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151527.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151527.hg.1 | ENST00000570223.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151527.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151527.hg.1 | TC0600007559.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600151528.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151528.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151528.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151528.hg.1 | ENST00000426185.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151528.hg.1 | ENST00000562344.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600151528.hg.1 | NR_003948.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600151528.hg.1 | TC0600007559.hg.1 | Clariom D TC |
