Assay Details
Gene Symbol: MIR577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030303.1 | 1 | 47 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | arylsulfatase family member J | ARSJ | -675.3kb | Anti-sense | Chr. 4 - 113899929 - 113980125 on NCBI Build GRCh38 | |
Downstream | N-deacetylase and N-sulfotransferase 4 | NDST4 |
N-HSST, N-HSST 4, NDST-4, NHSST4 |
170.9kb | Anti-sense | Chr. 4 - 114824122 - 115114098 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400008484.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000591.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525670 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525664 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525710 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525665 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525707 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525662 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525669 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366893 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525657 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15604026 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366884 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525701 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525681 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525713 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525700 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525687 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525668 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366898 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525689 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366857 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366861 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366880 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525705 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366900 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525698 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525662 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525670 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366884 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15604026 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525664 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525687 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525657 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366898 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525705 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366900 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525701 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366857 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525690 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525702 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525660 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525681 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525671 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525668 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525675 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525685 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525710 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366861 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525679 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525669 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525713 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525707 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525689 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366893 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525700 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03366880 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525698 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525704 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525674 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525686 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23525665 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400008484.hg.1 | ENST00000385196.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008484.hg.1 | NR_030303.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04008656.hg.1 | TC04000591.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04008656.hg.1 | ENST00000385196.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04008656.hg.1 | NR_030303.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400008484.hg.1 | ENST00000385196.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008484.hg.1 | NR_030303.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158262.hg.1 | ENST00000385196.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158262.hg.1 | NR_030303.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158262.hg.1 | ENST00000385196.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400158262.hg.1 | NR_030303.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400158262.hg.1 | TC0400008484.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3310870 | NR_030303.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3310870 | NR_030303.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3310870 | NR_030303.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3310870 | NR_030303.1 | human_transcriptome |
