Assay Details
Gene Symbol: MIR583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030309.1 | 1 | 43 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101929710 | LOC101929710 | Sense | Chr. 5 - 95962001 - 96631085 on NCBI Build GRCh38 | ||
Downstream | proprotein convertase subtilisin/kexin type 1 | PCSK1 |
BMIQ12, NEC1, PC1, PC3, SPC3 |
311.1kb | Anti-sense | Chr. 5 - 96390336 - 96433281 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500008140.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000466.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301558 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301546 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301563 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301555 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301573 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122298 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122316 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122274 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122319 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301579 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301541 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122264 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301528 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122300 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301571 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301569 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301533 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301574 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301543 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301562 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301539 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301561 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122278 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122301 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301559 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301551 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301578 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301537 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301527 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301579 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301569 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301543 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301546 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122298 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122301 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122274 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301527 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301558 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301562 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301539 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301545 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301573 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301537 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301568 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301578 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301541 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301540 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301551 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301561 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301563 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122273 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122300 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122316 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301555 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301571 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301528 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301559 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301538 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301553 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301574 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301533 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122278 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24301535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122264 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04122319 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05006599.hg.1 | TC05000466.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05006599.hg.1 | ENST00000384846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05006599.hg.1 | NR_030309.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500008140.hg.1 | ENST00000384846.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500008140.hg.1 | NR_030309.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500008140.hg.1 | ENST00000384846.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500008140.hg.1 | NR_030309.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500155718.hg.1 | ENST00000384846.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500155718.hg.1 | NR_030309.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500155718.hg.1 | ENST00000384846.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500155718.hg.1 | NR_030309.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500155718.hg.1 | TC0500008140.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3308200 | NR_030309.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3308200 | NR_030309.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308200 | NR_030309.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308200 | NR_030309.1 | human_transcriptome |
