Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007296.hg.1 |
ENST00000410758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007296.hg.1 |
ENST00000581232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007296.hg.1 |
ENST00000581232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007296.hg.1 |
ENST00000581232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007296.hg.1 |
ENST00000581232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007296.hg.1 |
NR_031720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002037.hg.1 |
ENST00000410758.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002037.hg.1 |
NR_031720.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002037.hg.1 |
TC18000174.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1800007296.hg.1 |
ENST00000410758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007296.hg.1 |
ENST00000581232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007296.hg.1 |
ENST00000581232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007296.hg.1 |
ENST00000581232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007296.hg.1 |
ENST00000581232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007296.hg.1 |
NR_031720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800149930.hg.1 |
ENST00000410758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149930.hg.1 |
ENST00000581232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149930.hg.1 |
NR_031720.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800149930.hg.1 |
TC1800007296.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800149930.hg.1 |
ENST00000410758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149930.hg.1 |
ENST00000581232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149930.hg.1 |
NR_031720.1 |
Refseq |