Assay Details
Gene Symbol: LINC01541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038325.1 | 3 - 4 | 328 | 110 | ||
GenBank mRNA | BX648957.1 | - | 3 - 4 | 329 | 110 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
Gilles de la Tourette syndrome chromosome region, candidate 1 (non-protein coding) |
GTSCR1 | -860.7kb | Sense | Chr. 18 - 70630519 - 70650857 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC102724913 | LOC102724913 | 198.3kb | Sense | Chr. 18 - 71732615 - 71782226 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000569.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18005988.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005988.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005988.hg.1 | TC18000569.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18005984.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005984.hg.1 | TC18000569.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18005987.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005987.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005987.hg.1 | TC18000569.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005985.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005985.hg.1 | TC18000569.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18005986.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005986.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005986.hg.1 | TC18000569.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800161735.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161735.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161735.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161735.hg.1 | TC1800008997.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800161735.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161735.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161735.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161736.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161736.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161736.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161736.hg.1 | TC1800008997.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800161736.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161736.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161736.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | TC1800008997.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161737.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | TC1800008997.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161738.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | TC1800008997.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800161739.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000566582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | ENST00000568095.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038326.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008997.hg.1 | NR_038326.1 | Refseq |
