Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7972933 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7972932 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
238127_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000611082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
ENST00000611082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004892.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13004892.hg.1 |
TC13000430.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000458001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000611082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
ENST00000611082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300010008.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167268.hg.1 |
ENST00000458001.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167268.hg.1 |
ENST00000611082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167268.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167268.hg.1 |
ENST00000458001.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167268.hg.1 |
ENST00000611082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167268.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167268.hg.1 |
TC1300010008.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300167267.hg.1 |
ENST00000458001.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167267.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167267.hg.1 |
ENST00000458001.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300167267.hg.1 |
NR_044995.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300167267.hg.1 |
TC1300010008.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
238127_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
238127_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
238127_at11 |
- |
- |
Human Genome U95 Set |
84444_PM_at |
- |
- |
Human Genome U95 Set |
84444_at |
- |
- |
Human Genome U133 Set |
238127_PM_at |
- |
- |
Human Genome U133 Set |
238127_at |
- |
- |