Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012817.hg.1 |
ENST00000432370.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012817.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012817.hg.1 |
TC10001034.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
ENST00000420634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
ENST00000420634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012819.hg.1 |
ENST00000432370.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012819.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012819.hg.1 |
TC10001034.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
ENST00000420634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
ENST00000420634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009781.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166791.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166791.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166791.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166791.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166791.hg.1 |
TC1000009781.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000166793.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166793.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166793.hg.1 |
ENST00000432370.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166793.hg.1 |
NR_046108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166793.hg.1 |
TC1000009781.hg.1 |
Clariom D TC |