Assay Details
Gene Symbol: LINC00659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046224.1 | 1 - 2 | 53 | 74 | ||
GenBank mRNA | DA633805.1 | - | 1 - 2 | 118 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neurotensin receptor 1 (high affinity) | NTSR1 | NTR | -12.5kb | Anti-sense | Chr. 20 - 62708770 - 62762771 on NCBI Build GRCh38 |
Downstream | MRG/MORF4L binding protein | MRGBP |
C20orf20, Eaf7, MRG15BP, URCC4 |
19.6kb | Anti-sense | Chr. 20 - 62796405 - 62801738 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
