Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000436929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000436929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000436929.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013185.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198395.hg.1 |
ENST00000436929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198395.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198395.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198395.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198395.hg.1 |
ENST00000436929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198395.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198395.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198395.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198395.hg.1 |
TC0300013185.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000436929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000436929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000436929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013185.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198394.hg.1 |
ENST00000436929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198394.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198394.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198394.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198394.hg.1 |
ENST00000436929.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198394.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198394.hg.1 |
ENST00000453180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300198394.hg.1 |
NR_046390.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300198394.hg.1 |
TC0300013185.hg.1 |
Clariom D TC |