Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012892.hg.1 |
ENST00000467794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012892.hg.1 |
ENST00000467794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012892.hg.1 |
ENST00000467794.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012892.hg.1 |
NR_046618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012892.hg.1 |
NR_046618.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012892.hg.1 |
NR_046618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196372.hg.1 |
ENST00000467794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196372.hg.1 |
NR_046618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196372.hg.1 |
ENST00000467794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196372.hg.1 |
NR_046618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196372.hg.1 |
TC0300012892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300196373.hg.1 |
ENST00000467794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196373.hg.1 |
NR_046618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196373.hg.1 |
ENST00000467794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196373.hg.1 |
NR_046618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196373.hg.1 |
TC0300012892.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012892.hg.1 |
ENST00000467794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012892.hg.1 |
ENST00000467794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012892.hg.1 |
ENST00000467794.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012892.hg.1 |
NR_046618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012892.hg.1 |
NR_046618.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012892.hg.1 |
NR_046618.2 |
Refseq |