Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010804.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010804.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010804.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010804.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010804.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010804.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010804.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010804.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700176984.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700176984.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700176984.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700176984.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700176984.hg.1 |
TC0700010804.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700176983.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700176983.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700176983.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700176983.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700176983.hg.1 |
TC0700010804.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700010804.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010804.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010804.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010804.hg.1 |
ENST00000433519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010804.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010804.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010804.hg.1 |
NR_046711.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010804.hg.1 |
NR_046711.1 |
Refseq |