Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011131.hg.1 |
ENST00000442411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011131.hg.1 |
ENST00000442411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011131.hg.1 |
NR_047551.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011131.hg.1 |
NR_047551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179574.hg.1 |
ENST00000442411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179574.hg.1 |
NR_047551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179574.hg.1 |
ENST00000442411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179574.hg.1 |
NR_047551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179574.hg.1 |
TC0700011131.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011131.hg.1 |
ENST00000442411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011131.hg.1 |
ENST00000442411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011131.hg.1 |
NR_047551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011131.hg.1 |
NR_047551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179572.hg.1 |
ENST00000442411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179572.hg.1 |
NR_047551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179572.hg.1 |
ENST00000442411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700179572.hg.1 |
NR_047551.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700179572.hg.1 |
TC0700011131.hg.1 |
Clariom D TC |