Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152482.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152482.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152482.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152482.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152482.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152482.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152482.hg.1 |
TC1100007422.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100152480.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152480.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152480.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152480.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152480.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152480.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152480.hg.1 |
TC1100007422.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100152479.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152479.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152479.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152479.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152479.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152479.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152479.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152479.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152479.hg.1 |
TC1100007422.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100152481.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152481.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152481.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152481.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152481.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100152481.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100152481.hg.1 |
TC1100007422.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000529769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000530051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000532307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
ENST00000614989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007422.hg.1 |
NR_047569.3 |
Refseq |