| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178488.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178488.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178488.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178488.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178488.hg.1 |
TC1700010013.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010013.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178483.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178483.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178483.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178483.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178483.hg.1 |
TC1700010013.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700178487.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178487.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178487.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178487.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178487.hg.1 |
TC1700010013.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700178482.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178482.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178482.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178482.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178482.hg.1 |
TC1700010013.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700178486.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178486.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178486.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178486.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178486.hg.1 |
TC1700010013.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700178484.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178484.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178484.hg.1 |
ENST00000451099.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700178484.hg.1 |
NR_048576.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700178484.hg.1 |
TC1700010013.hg.1 |
Clariom D TC |