Assay Details
Gene Symbol: LOC100130476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049793.1 | 3 - 4 | 1052 | 89 | ||
GenBank mRNA | AK124173.1 | - | 3 - 4 | 1052 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100507406 | LOC100507406 | -84.6kb | Sense | Chr. 6 - 137730170 - 137739037 on NCBI Build GRCh38 | |
Downstream | TNF alpha induced protein 3 | TNFAIP3 |
A20, AISBL, OTUD7C, TNFA1P2 |
997.0 | Anti-sense | Chr. 6 - 137866317 - 137883314 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3365352 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3365352 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192140.hg.1 | ENST00000440578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192140.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192140.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192140.hg.1 | ENST00000440578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192140.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192140.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192140.hg.1 | TC0600013316.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600192139.hg.1 | ENST00000440578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192139.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192139.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192139.hg.1 | ENST00000440578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192139.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600192139.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600192139.hg.1 | TC0600013316.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000431144.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000440578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000448942.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606327.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000606998.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | ENST00000607671.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013316.hg.1 | NR_049793.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 243871_at | - | - |
Human Genome U133 Plus 2.0 Array | 243871_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 243871_at8 | - | - |
Human Genome U95 Set | 60908_PM_at | - | - |
Human Genome U95 Set | 60908_at | - | - |
Human Genome U133 Set | 243871_at | - | - |
Human Genome U133 Set | 243871_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3257004 | BC041790.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3257004 | BC041790.1 | human_transcriptome |
