Assay Details
Gene Symbol: LINC00534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_051989.1 | 2 - 3 | 354 | 92 | ||
GenBank mRNA | BG492574.1 | - | 2 - 3 | 305 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | calbindin 1 | CALB1 |
CALB, D-28K |
-163.3kb | Anti-sense | Chr. 8 - 90058608 - 90082881 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1030 | LINC01030 | 241.1kb | Anti-sense | Chr. 8 - 90592775 - 90606063 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156236.hg.1 | TC0800008186.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800156235.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156235.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156235.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800156235.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800156235.hg.1 | TC0800008186.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000517400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000523406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | ENST00000524361.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | NR_051989.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008186.hg.1 | NR_051989.1 | Refseq |
