Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010363.hg.1 |
ENST00000452497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010363.hg.1 |
ENST00000452497.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010363.hg.1 |
NR_051997.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010363.hg.1 |
NR_051997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170492.hg.1 |
NR_051997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170492.hg.1 |
NR_051997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170492.hg.1 |
TC1000010363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000010363.hg.1 |
ENST00000452497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010363.hg.1 |
ENST00000452497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010363.hg.1 |
NR_051997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010363.hg.1 |
NR_051997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170493.hg.1 |
ENST00000452497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170493.hg.1 |
NR_051997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170493.hg.1 |
ENST00000452497.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000170493.hg.1 |
NR_051997.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000170493.hg.1 |
TC1000010363.hg.1 |
Clariom D TC |