Assay Details
| Gene Symbol: SLC16A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270622.1 | NP_001257551.1 | 4 - 5 | 664 | 90 | |
| NM_001270623.1 | NP_001257552.1 | 4 - 5 | 684 | 90 | ||
| NM_004731.4 | NP_004722.2 | 3 - 4 | 522 | 90 | ||
| NR_073055.1 | 5 - 6 | 798 | 90 | |||
| NR_073056.1 | 5 - 6 | 659 | 90 | |||
| XM_005269231.4 | XP_005269288.1 | 3 - 4 | 458 | 90 | ||
| XM_011538989.2 | XP_011537291.1 | 5 - 6 | 868 | 90 | ||
| XM_011538990.2 | XP_011537292.1 | 4 - 5 | 760 | 90 | ||
| XM_011538991.2 | XP_011537293.1 | 4 - 5 | 558 | 90 | ||
| XM_011538992.2 | XP_011537294.1 | 4 - 5 | 1493 | 90 | ||
| XM_011538993.2 | XP_011537295.1 | 6 - 7 | 1258 | 90 | ||
| XM_011538995.2 | XP_011537297.1 | 6 - 7 | 1253 | 90 | ||
| XM_017020224.1 | XP_016875713.1 | 7 - 8 | 1179 | 90 | ||
| XM_017020225.1 | XP_016875714.1 | 7 - 8 | 1172 | 90 | ||
| XM_017020226.1 | XP_016875715.1 | 7 - 8 | 1307 | 90 | ||
| XM_017020227.1 | XP_016875716.1 | 6 - 7 | 853 | 90 | ||
| GenBank mRNA | AF049608.1 | - | 4 - 5 | 519 | 90 | |
| AF058056.1 | - | 3 - 4 | 485 | 90 | ||
| AK301725.1 | - | 5 - 6 | 786 | 90 | ||
| AK310281.1 | - | 5 - 6 | 626 | 90 | ||
| AK313345.1 | - | 3 - 4 | 492 | 90 | ||
| BC030693.2 | - | 3 - 4 | 514 | 4827290 | 90 | |
More Information
Set Membership: |
|

SLC16A7
Genomic Map