Assay Details
| Gene Symbol: TFAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073073.1 | 2 - 3 | 626 | 120 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin conjugating enzyme E2 D1 | UBE2D1 |
E2(17)KB1, SFT, UBC4/5, UBCH5, UBCH5A |
-15.5kb | Sense | Chr. 10 - 58334979 - 58370753 on NCBI Build GRCh38 |
| Downstream | BicC family RNA binding protein 1 | BICC1 |
BICC, CYSRD |
126.2kb | Sense | Chr. 10 - 58512970 - 58831437 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
DNA-dependent DNA replication
regulation of transcription from RNA polymerase I promoter
transcription from RNA polymerase II promoter
transcription from mitochondrial promoter
transcription initiation from mitochondrial promoter
mitochondrion organization
mitochondrial respiratory chain complex assembly
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription coactivator binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
DNA binding, bending
heat shock protein binding
poly(A) RNA binding
mitochondrial light strand promoter sense binding
DNA-dependent DNA replication
regulation of transcription from RNA polymerase I promoter
transcription from RNA polymerase II promoter
transcription from mitochondrial promoter
transcription initiation from mitochondrial promoter
mitochondrion organization
mitochondrial respiratory chain complex assembly
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription coactivator binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
DNA binding, bending
heat shock protein binding
poly(A) RNA binding
mitochondrial light strand promoter sense binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000395377.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | ENST00000487519.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_001270782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NM_003201.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007701.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152331.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152331.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152331.hg.1 | ENST00000373899.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000152331.hg.1 | NR_073073.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000152331.hg.1 | TC1000007701.hg.1 | Clariom D TC |

TFAM
Genomic Map