Assay Details
| Gene Symbol: TFAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073073.1 | 3 - 4 | 686 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin conjugating enzyme E2 D1 | UBE2D1 |
E2(17)KB1, SFT, UBC4/5, UBCH5, UBCH5A |
-15.9kb | Sense | Chr. 10 - 58334979 - 58370753 on NCBI Build GRCh38 |
| Downstream | BicC family RNA binding protein 1 | BICC1 |
BICC, CYSRD |
124.7kb | Sense | Chr. 10 - 58512970 - 58831437 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

TFAM
Genomic Map